Please login to your online account to display your discounted pricing

Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems


Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids

Manufacturer: hoefer™  SE250

 View more versions of this product

Catalog No. 03-500-483

  • / Each
 

Description & Specifications

Specifications

Wattage 12w
For Use With (Equipment) Precast gel or order two SE 211A combs, two sets of 8cm long T-shape spacers, 10 x 8cm rectangular Glass and notched Alumina plates and a gel caster
Depth (English) 6.29 in.
Temperature (Metric) Storage 15° to 30°C
Includes Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, two 10 x 8cm notched Alumina plates, four spring clamps, gel seal, well locating decal and spacer mate
Length (English) Gel 3.14 in.
Humidity Range Up to 80%
Length (Metric) Gel 8cm
Volume (English) Buffer 10.14 oz.
Width (Metric) Exterior 16.5cm
Height (English) Exterior 6.29 in.
Width (Metric) Gel 10cm
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Width (English) Exterior 6.49 in.
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Depth (Metric) 16cm
Voltage 500V
Amperage 500mA
Width (English) Gel 3.93 in.
Height (Metric) Exterior 16cm
Volume (Metric) Buffer 300mL