Please login to your online account to display your discounted pricing

Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems

Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids

$1,329.62 - $1,329.62

Specifications

Amperage 500mA
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Depth (English) 6.29 in.
Depth (Metric) 16cm
Volume (English) Buffer 10.14 oz.
View More Specs
Includes:  SE250 Basic System includes: upper buffer chamber, lower buffer chamber, interlocking safety lid with leads, ten 10 x 8cm glass plates, two 10 x 8cm notched alumina plates, four spring clamps, gel seal, well locating decal, and Spacer Mate. SE250 Complete System also includes: Dual Gel Caster, two 0.75mm thick 10-well combs, and two pairs 0.75mm thick spacers. SE260 Basic System includes same items as SE250 Basic Systems, except the glass and alumina plates.SE260 Complete Systems add Dual Gel Caster, ten 10.5 x 10cm glass plates, five 10.5 x 10cm notched alumina plates, and two 10-well combs and two pairs spacers (either 0.75mm or 1.5mm thick; see ordering table).
Products
Catalog Number Mfr. No. Includes Height (Metric) Exterior Length (Metric) Gel Price Quantity    

03-500-483

 
hoefer™
SE250
Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, two 10 x 8cm notched Alumina plates, four spring clamps, gel seal, well locating decal and spacer mate 16cm 8cm Each for $1,329.62

03-500-490

 
hoefer™
SE260B
Upper Buffer chamber, lower Buffer chamber, interlocking safety lid with Leads, ten 10 x 8cm Glass plates, four spring clamps, gel seal, well locating decal and spacer mate 18cm 8/10.5cm Each for $1,329.62

03-500-111

 
hoefer™
SE260-10A-.75
Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers, well locating decals, gel seal 18cm 8/10.5cm Each for $1,329.62

SE26010A15

 
hoefer™
SE26010A15
Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers, well locating decals, gel seal 18cm 8/10.5cm Each for $1,329.62

03-500-110

 
hoefer™
SE25010A75
Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers, well locating decals, gel seal 16cm 8cm Each for $1,329.62
Description & Specifications

Specifications

Amperage 500mA
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
Depth (English) 6.29 in.
Depth (Metric) 16cm
Volume (English) Buffer 10.14 oz.
Volume (Metric) Buffer 300mL
Width (English) Exterior 6.49 in.
Width (English) Gel 3.93 in.
Width (Metric) Exterior 16.5cm
Width (Metric) Gel 10cm
Humidity Range Up to 80%
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15° to 30°C
Voltage 500V
Wattage 12w

  • Run two gels at once
  • Produce high-resolution separations in as little as 45 minutes with no band “smiling”
  • Use alumina plates (included with specified models) for efficient cooling
  • Buffer volume: 300mL

Applications:

Ideal for uniform and gradient acrylamide, agarose, IEF, field inversion agarose and 2-D electrophoresis

CE Marked, CSA C22.2 1010.1, EN61010-1, UL3101-1.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.