Please login to your online account to display your discounted pricing

Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems


Combine the advantages of a small format and efficient cooling for rapid screening of protein and nucleic acids

Manufacturer: hoefer™  SE260-10A-.75

 View more versions of this product

Catalog No. 03-500-111

  • / Each
 

Description & Specifications

Specifications

Amperage 500mA
Depth (English) 6.29 in.
Depth (Metric) 16cm
Humidity Range Up to 80%
Includes Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, combs, 10-well, T-shape spacers, well locating decals, gel seal
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Temperature (Metric) Storage 15° to 30°C
Thickness (English) Spacer 0.2 in.
Thickness (Metric) Spacer 0.07cm
Voltage 500V
Wattage 12w
Volume (English) Buffer 10.14 oz.
Volume (Metric) Buffer 300mL
Length (English) Gel 3.14/4.13 in.
Length (Metric) Gel 8/10.5cm
Width (English) Gel 3.93 in.
Width (Metric) Gel 10cm
Height (English) Exterior 7.08 in.
Height (Metric) Exterior 18cm
Width (English) Exterior 6.49 in.
Width (Metric) Exterior 16.5cm
Certifications/Compliance EN61010-1, UL3101-1, CSA C22.2 1010.1, CE