Hoefer™ Maxi-Plus and Maxi-Plus Submarine Gel Electrophoresis Units
Ideal for high resolution analytical and preparative studies of nucleic acids
Manufacturer:  Hoefer™ SUB25C
 View more versions of this product
Catalog No. 03-500-793
Specifications
Specifications
500mA
Up to 80%
624
45°C
500w
101.4 oz.
11.8 in.
9.8 in.
3.5 in.
22 in.
13.7 in.
EN61010-1, CE
22 x 13.7 x 3.5 in. (56 x 35 x 9cm)
Buffer chamber assembly with cooling ports, safety lid with high voltage Leads, running tray with casting gates, 1mm thick, 16-well combs, colored loading strips, Buffer recirculation ports
113°F
1000V
Submarine gel electrophoresis unit
3000mL
30cm
25cm
9cm
56cm
35cm
Documents