Please login to your online account to display your discounted pricing

Hoefer™ HE-PLUS Horizontal Electrophoresis System


Compact, easy-to-use system completely integrated with a built-in variable power supply

Manufacturer: hoefer™  HEPLUS230V

 View more versions of this product

Catalog No. 03-500-554

  • / Each
 

Description & Specifications

Specifications

Sample Size 112
Length (Metric) Gel 13cm
Length (English) Exterior 6.6 in.
Width (English) Exterior 9.6 in.
Height (English) Exterior 2.4 in.
Volume (Metric) Buffer 350mL
Certifications/Compliance CE, CSA, TVE
Includes Gel tank, safety lid, power supply, large gel tray, medium gel trays, large or medium comb 14, 28-well, casting stand
Product Type Horizontal electrophoresis system
Hertz 50/60Hz
Length (Metric) Exterior 17cm
Wattage 45w
Width (Metric) Gel 12.5cm
Width (English) Gel 4.9 in.
Display Type LED
Electrical Requirements 230V 50/60Hz
Weight (Metric) 410g
Timer 99.59 Hours or continuous run
Voltage 230V
Amperage 10 to 400mA
Dimensions (L x W x H) Exterior 6.6 x 9.6 x 2.4 in. (17 x 24.5 x 6.2cm)
Weight (English) 0.9 lb.
Volume (English) Buffer 11.8 oz.
Length (English) Gel 5.1 in.
Width (Metric) Exterior 24.5cm
Height (Metric) Exterior 6.2cm