Hoefer™ HE-PLUS Casting Kits
For use with HE-PLUS Horizontal Electrophoresis System
Manufacturer:  Hoefer™ HEPLUS16
 View more versions of this product
Catalog No. 03-500-560
Specifications Description & Specifications
Specifications
Mini Casting Kit
HE-plus horizontal electrophoresis system
Four gel trays (6 x 6cm), two combs (5/8-well) and casting stand
6 x 6cm