Hoefer™ HE 99X Max Submarine Electrophoresis Unit
Offers no-mess gel casting without tape or dams
Manufacturer:  Hoefer™ HE99X
 View more versions of this product
Catalog No. 36-100-3898
Includes: Buffer chamber with electrodes and leveling feet, safety lid with leads, bubble level. Complete system (03-500-121) also includes 15 × 20cm gel casting tray and UV-transparent running tray; 1.5mm thick, 15-well comb; adjustable comb back that permits control of the depth of the sample wells
Description
Description
  • Unit accommodates a variety of gel lengths for maximum resolution of DNA fragments
  • Agarose bridges are also accommodated, so gels can be run in nonsubmarine fashion
  • Rugged design provides reliability and safety
Specifications
Specifications
100 mA
14.1 x 7.1 x 5.5 in. (36 x 18.2 x 14 cm)
5.5 in.
Up to 80%
14.1 in.
36 cm
60
45°C
40.5 oz.
20 w
5.9 in.
15 cm
EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Horizontal Agarose Electrophoresis Unit
14 cm
Buffer chamber assembly, safety lid with high voltage Leads, Foam gaskets, bubble level
5.9, 3.9 or 7.8 in.
15, 10 or 20 cm
113°F
200 V
1200 mL
7.1 in.
18.2 cm