missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hoefer™ Gel Casting Accessories for HE 99X Max Submarine Units: Running Trays

For use with Hoefer HE 99X Max Submarine Electrophoresis Units

$275.20 - $278.07

Specifications

Color UV Transparent
Product Type Running Tray
Material Acrylic
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (English) Length (Metric) Width (English) Width (Metric) Price Quantity  
03-500-147 Hoefer™
HE92X-10
5.9 in. 15cm 3.93 in. 10cm Each for $275.20
03-500-149 Hoefer™
HE92X20
7.8 in. 20cm 5.9 in. 15cm Each for $278.07
Specifications

Description

  • Running trays are UV-transparent acrylic

Description & Specifications

Specifications

UV Transparent
Running Tray
Acrylic