Hoefer™ Gel Casting Accessories for HE 99X Max Submarine Units: Casting Kits
For use with Hoefer HE 99X Max Submarine Electrophoresis Units
Manufacturer:  Hoefer™ HE97X10
 View more versions of this product
Catalog No. 36-100-3901
Description
Description
For use with Hoefer HE 99X Max Submarine Electrophoresis Units
  • Casting kits include necessary components to cast agarose gels without tape—includes casting tray, running tray, and foam gaskets
Specifications
Specifications
5.9 x 3.9 in. (15 x 10 cm)
Casting Kit
Running tray, casting tray, Foam gasket
15 cm
10 cm
HE99X Max Submarine Electrophoresis Unit
15 x 10 cm
5.9 in.
3.9 in.