Cytiva (Formerly GE Healthcare Life Sciences) Gel Casters for Mini Vertical Electrophoresis Systems
Easy-to-use casters
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) SE275
 View more versions of this product
Catalog No. 36-100-3989
Description
Description
SE245 Dual-Gel Caster
  • For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
  • SE235 4-Gel Caster, SE275 4-Gel Caster, and SE215 10-Gel Caster
    • Suitable for single concentration and gradient gels
    • SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
    • SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
    Specifications
    Specifications
    SE 250 Mini Vertical Gel Electrophoresis Unit
    Gel Caster
    Casting cradle with gasket, face, Glass, alumina, space saver plates, filler sheets, plugs
    1 to 4