Cytiva (Formerly GE Healthcare Life Sciences) Gel Casters for Mini Vertical Electrophoresis Systems
Easy-to-use casters
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) SE245
 View more versions of this product
Catalog No. 36-100-3996
Description
Description
SE245 Dual-Gel Caster
  • For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
  • SE235 4-Gel Caster, SE275 4-Gel Caster, and SE215 10-Gel Caster
    • Suitable for single concentration and gradient gels
    • SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
    • SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
    Specifications
    Specifications
    SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
    Gel Caster
    Casting cradle, clamp assemblies, sealing gasket set, cams
    1 to 2
    Documents