Please login to your online account to display your discounted pricing

Hoefer™ Combs for SUB25 Maxi-Plus Submarine Gel Electrophoresis Unit


Manufacturer: hoefer™  SUB25C252MC

 View more versions of this product

Catalog No. 03-500-799

  • / Each
 

Description & Specifications

Specifications

Thickness (English) 0.07 in.
For Use With (Equipment) SUB25 Maxi-Plus unit
Width (English) Gel 9.84 in.
Volume (Metric) 25μL
Volume (English) 0.0008 oz.
For Use With (Application) 52 sample throughput (multi-channel pipette compatible)
Thickness (Metric) 2mm