Please login to your online account to display your discounted pricing

Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units


Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Manufacturer: hoefer™  SE5111210

 View more versions of this product

Catalog No. 03-500-237

  • / Each
 

Description & Specifications

Specifications

Product Type Comb
Thickness (English) 0.03 in.
No. of Wells 12
Depth (Metric) 2.5cm
For Use With (Equipment) SE 600, SE 400 series vertical electrophoresis system
Depth (English) 0.98 in.
Width (Metric) 0.76cm
Volume (Metric) Well 7.7μL/mm
Material PTFE
Thickness (Metric) 0.1cm