Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Manufacturer:  Hoefer™ SE5112015
 View more versions of this product
Catalog No. 03-500-193
Description
Description
  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25mm, except for the 28-well preparative combs which produce well depths of 15mm
  • Adjustable comb back (available separately, below) converts 25mm deep combs to 10 or 15mm depth
Specifications
Specifications
0.05 in.
0.41cm
0.98 in.
SE 600, SE 400 series vertical electrophoresis system
20
0.15cm
6.2μL/mm
2.5cm
Comb
PTFE
Documents