missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Manufacturer:  Cytiva SE511101.0

 View more versions of this product

Catalog No. 36-100-3866



Description

Description

  • Lightweight
  • Virtually unbreakable

  • Come attached to a standard back which produces maximum well depths of 25mm, except for the 28-well preparative combs which produce well depths of 15mm
  • Adjustable comb back (available separately, below) converts 25mm deep combs to 10 or 15mm depth

Specifications

Specifications

0.98 in.
SE 600, SE 400 Series Vertical Electrophoresis System
PTFE
0.03 in.
8.3μL/ mm
2.5 cm
Comb
10
0.1 cm
0.83 cm
SDS
Documents