missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Manufacturer: Cytiva SE511281.5
Description
- Lightweight
- Virtually unbreakable
- Come attached to a standard back which produces maximum well depths of 25mm, except for the 28-well preparative combs which produce well depths of 15mm
- Adjustable comb back (available separately, below) converts 25mm deep combs to 10 or 15mm depth
Specifications
0.98 in. | |
SE 600, SE 400 Series Vertical Electrophoresis System | |
PTFE | |
0.05 in. | |
4.1μL/ mm |
2.5 cm | |
Comb | |
28 | |
0.15 cm | |
0.27 cm |