Hoefer™ Combs for SE 600 Series or SE 400 Series Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Manufacturer:  Hoefer™ SE511DR15
 View more versions of this product
Catalog No. 03-500-200
Description
Description
  • Lightweight
  • Virtually unbreakable
  • Come attached to a standard back which produces maximum well depths of 25mm, except for the 28-well preparative combs which produce well depths of 15mm
  • Adjustable comb back (available separately, below) converts 25mm deep combs to 10 or 15mm depth
Specifications
Specifications
0.05 in.
171μL/mm
2.5cm
Comb
PTFE
11.3cm
0.15cm
0.98 in.
SE 600, SE 400 series vertical electrophoresis system
1 Preparative, 2 Reference
4.4 in.
Documents