Cytiva (Formerly GE Healthcare Life Sciences) Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) SE211A51.5
 View more versions of this product
Catalog No. 36-100-3880
Description
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
  • Lightweight
  • Virtually unbreakable
Specifications
Specifications
0.51 in.
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
PTFE
0.05 in.
19.5μL/ mm
1.3 cm
Comb
5
0.15 cm
1.3 cm