Please login to your online account to display your discounted pricing

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units


Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

Manufacturer: hoefer™  SE211A101.5

 View more versions of this product

Catalog No. 03-500-165

  • / Each
 

Description & Specifications

Specifications

Thickness (English) 0.05 in.
Thickness (Metric) 0.15cm
Width (Metric) 0.48cm
Volume (Metric) Well 7.2μL/mm
Depth (English) 0.51 in.
Depth (Metric) 1.3cm
For Use With (Equipment) SE 250, SE 260, MiniVE™ vertical electrophoresis system
Product Type Comb
No. of Wells 10
Material PTFE