Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Manufacturer:  Hoefer™ HE91BK
 View more versions of this product
Catalog No. 36-100-3923
Requires : Comb back must be ordered separately (see ordering table)
Description
Description
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
Combs can be used interchangeably with a single comb back.
Specifications
Specifications
0.19 in.
HE99X Max Submarine Electrophoresis Unit
2 Screws
0.5 cm
Comb Back