Please login to your online account to display your discounted pricing
Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit
Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit
$101.71 - $129.15
Specifications
Depth (English) Pan 0.19 in.
Depth (Metric) Pan 0.5cm
For Use With (Equipment) HE99X max submarine electrophoresis unit
View More Specs
Requires:  Comb back must be ordered separately (see ordering table)
Products
Catalog Number Mfr. No. No. of Wells Product Type Thickness (Metric) Price Quantity    
36-100-3923
hoefer™
HE91BK
Each for $101.71
03-500-143
hoefer™
HE91A3010
30 Backless Comb 0.1cm Each for $129.15
36-100-3930
hoefer™
HE91A201.5
20 Backless Comb 0.15cm Each for $125.16
03-500-138
hoefer™
HE91A151.5
15 Backless Comb 0.15cm Each for $129.15
Description & Specifications
Specifications
Depth (English) Pan 0.19 in.
Depth (Metric) Pan 0.5cm
For Use With (Equipment) HE99X max submarine electrophoresis unit
Combs can be used interchangeably with a single comb back.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.