missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units

For use with Mighty Small electrophoresis units

Manufacturer:  Hoefer™ SE255D

 View more versions of this product

Catalog No. 03-500-488


 

Specifications Description & Specifications

Specifications

SE 260
Buffer Chamber