missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units

For use with Mighty Small electrophoresis units

Manufacturer:  Hoefer™ SE254B

 View more versions of this product

Catalog No. 03-500-486


 

Specifications Description & Specifications

Specifications

SE 250 or SE 260
Buffer Chamber
Foam gasket