missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hoefer™ Buffer Chambers for Mighty Small™ Electrophoresis Units

For use with Mighty Small electrophoresis units

$396.40 - $1,174.31

Specifications

Product Type Buffer Chamber
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
03-500-488 Hoefer™
SE255D
SE 260 Each for $396.40
03-500-486 Hoefer™
SE254B
SE 250 or SE 260 Each for $1,174.31
Specifications Description & Specifications

Specifications

Buffer Chamber