Please login to your online account to display your discounted pricing
Thermo Scientific™ Microm™ Cryostats Heat Extractors
Use Thermo Scientific™ Microm™ Cryostats Heat Extractors with Richard-Allan Scientific™ Cryostats.
Manufacturer: thermo scientific™  524490
 View more versions of this product
Catalog No. MIC524490
  • / Each
Description & Specifications
Specifications
Description Heat Extractor
For Use With HM 520, HM 525
Item Description Heat Extractor
Use Thermo Scientific™ Microm™ Cryostats Heat Extractors with Richard-Allan Scientific™ Cryostats.
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier