Learn More
HLA-G Monoclonal Antibody (87G), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 46995742
Description
Description: The monoclonal antibody 87G recognizes human HLA-G, a member of the Human Leukocyte Antigen family but as part of the nonclassical MHC type involved in inhibiting immune reponses. HLA-G has seven reported isoforms. The antibody 87G recognizes both HLA-G1 and the solube HLA-G5. Expression of HLA-G is found primarily in fetal trophoblast cells as they invade the maternal decidua thereby protecting the fetus from the maternal immune system. Like the highly mitotic trophoblast, abundant HLA-G protein expression has been identified in some tumors, including melanoma, breast carcinoma and renal carcinoma as well as CLL, AML and B-CLL. Some expression has also been found in pancreatic islets, erythroid and endothelial progenitors and the adult thymic medulla.HLA-G+ CD4 or CD8 cells have been identified in normal human peripheral blood and are thought to act as regulatory cells in that they are hypoproliferative with a unique cytokine profile differing from Tregs. The receptors for HLA-G are CD85j/ILT2, CD85d/ILT4, and CD158. Recent studies have shown a role for HLA-G in tolerance and maintece of transplanted organs. Applications Reported: This 87G antibody has been reported for use in flow cytometric analysis. Applications Tested: This 87G antibody has been pre-titrated and tested by flow cytometric analysis of stimulated monocytic cell line. This can be used at 5 μL (0.125 μg) per test.
HLA-G Monoclonal antibody specifically detects HLA-G in Human samples. It is validated for Flow Cytometry.Specifications
HLA-G | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P17693 | |
HLA-G | |
Affinity chromatography | |
RUO | |
3135 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
87G | |
PerCP-eFluor 710 | |
HLA-G | |
acrosomal protein SP-10; Acrosomal vesicle protein 1; ACRV1; b2 microglobulin; D11S4365; DADB-15K14.8; HLA class I histocompatibility antigen, alpha chain G; HLA class I molecule; HLA G antigen; HLA-6.0; HLAG; HLA-G; HLA-G histocompatibility antigen, class I, G; LOC768350; major histocompatibility complex, class I, G; MHC class I antigen G; MHC-G; MHCIY; MHC-like class I Y; sHLA-G; Soluble HLA class I histocompatibility antigen, alpha chain G; SP-10; SPACA2; sperm protein 10 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
For Research Use Only.