Please login to your online account to display your discounted pricing

HLA-E Mouse anti-Human, Clone: 3D12HLA-E, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $288.55

Specifications

Antigen HLA-E
Clone 3D12HLA-E
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013915

 
life technologies
LS14995380
25μg Each for $158.05

5013916

 
life technologies
LS14995382
100μg Each for $288.55
Description & Specifications

Specifications

Antigen HLA-E
Clone 3D12HLA-E
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 3D12 (HLA-E) recognizes human HLA-E, a member of the Human Leukocyte Antigen family. HLA-E is a MHC Class IB member, which has a specialized role in NK cell recognition. Binding of a restricted set of peptides derived from other Class I leader sequences to the HLA-E and B2M (beta2 microglobulin) complex leads to interaction with NKG2A on NK cells. This binding protects the HLA-G-expressing cell from NK cell killing. HLA-E expression is ubiquitous.