Please login to your online account to display your discounted pricing

HLA-DR Mouse anti-Human, PerCP-Cy5.5, Clone: LN3, eBioscience™

Mouse Monoclonal Antibody

$245.05 - $487.20

Specifications

Antigen HLA-DR
Clone LN3
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015886

 
life technologies
LS45995642
100 tests Each for $487.20

50245846

 
life technologies
LS45995641
25 tests Each for $245.05
Description & Specifications

Specifications

Antigen HLA-DR
Clone LN3
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-Cy5.5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.015μg)/test

The LN3 mAb reacts with the human major histocompatibility complex (MHC) class II, HLA-DR. HLA-DR is expressed on the surface of human antigen presenting cells (APC) including B cells, monocytes, macrophages, DCs, and activated T cells. HLA-DR is a heterodimeric transmembrane protein composed of α and β subunits and plays an important role in the presentation of peptides to CD4+ T lymphocytes.