missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HLA-DR Mouse anti-Human, PE-Cy5, Clone: LN3, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $341.88

Specifications

Antigen HLA-DR
Clone LN3
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014160 Life Technologies
LS15995641
25 Tests Each for $170.28
5014161 Life Technologies
LS15995642
100 Tests Each for $341.88
Specifications

Description

The LN3 mAb reacts with the human major histocompatibility complex (MHC) class II, HLA-DR. HLA-DR is expressed on the surface of human antigen presenting cells (APC) including B cells, monocytes, macrophages, DCs, and activated T cells. HLA-DR is a heterodimeric transmembrane protein composed of α and β subunits and plays an important role in the presentation of peptides to CD4+ T lymphocytes.
Description & Specifications

Specifications

HLA-DR
Murine
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.03μg)/test
LN3
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer