Please login to your online account to display your discounted pricing

HLA-DR Mouse anti-Human, PE, Clone: L243, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $361.05

Specifications

Antigen HLA-DR
Clone L243
Host Species Mouse
Species Reactivity Canine, Cynomolgus, Human, Non-human primate, Rhesus Ma
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011312

 
affymetrix
12995241
25 tests Each for $129.05

5011313

 
affymetrix
12995242
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen HLA-DR
Clone L243
Host Species Mouse
Species Reactivity Canine, Cynomolgus, Human, Non-human primate, Rhesus Ma
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The L243 mAb reacts with the human major histocompatibility complex (MHC) class II, HLA-DR. HLA-DR is expressed on the surface of human antigen presenting cells (APC) including B cells, monocytes, macrophages, DCs, and activated T cells. HLA-DR is a heterodimeric transmembrane protein composed of alpha and beta subunits and plays an important role in the presentation of peptides to CD4+ T lymphocytes. The L243 antibody recognizes a different epitope than the LN3 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes.