Please login to your online account to display your discounted pricing

HLA-DR Mouse anti-Human, Clone: LN3, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $158.05

Specifications

Antigen HLA-DR
Clone LN3
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013917

 
life technologies
LS14995680
25μg Each for $85.55

5013918

 
life technologies
LS14995682
100μg Each for $158.05
Description & Specifications

Specifications

Antigen HLA-DR
Clone LN3
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The LN3 mAb reacts with the human major histocompatibility complex (MHC) class II, HLA-DR. HLA-DR is expressed on the surface of human antigen presenting cells (APC) including B cells, monocytes, macrophages, DCs, and activated T cells. HLA-DR is a heterodimeric transmembrane protein composed of α and β subunits and plays an important role in the presentation of peptides to CD4+ T lymphocytes.