Please login to your online account to display your discounted pricing

HLA-ABC Mouse anti-Human, Clone: W6/32, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $143.55

Specifications

Antigen HLA-ABC
Clone W6/32
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122898

 
life technologies
LS14998380
25μg Each for $71.05

5013962

 
life technologies
LS14998382
100μg Each for $143.55
Description & Specifications

Specifications

Antigen HLA-ABC
Clone W6/32
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The W6/32 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I, HLA-A, B, C. MHC class I antigens associated with beta 2-microglobulin are expressed by all human nucleated cells and are central in cell-mediated immune response and tumor surveillance. W6/32 mAb recognizes a non-polymorphic epitope shared among products of the HLA-A, B, and C loci and immunoprecipitates both 43kDa and 11-12kDa chains. Crossreactivity is also seen in baboon, rhesus and cynomolgus monkey.