missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HLA-ABC Mouse anti-Human, Biotin, Clone: W6/32, eBioscience™

Mouse Monoclonal Antibody

$104.28 - $236.28

Specifications

Antigen HLA-ABC
Clone W6/32
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012305 Life Technologies
LS13998380
25μg Each for $104.28
501128896 Life Technologies
LS13998382
100μg Each for $236.28
Specifications

Description

The W6/32 monoclonal antibody reacts with the human major histocompatibility complex (MHC) class I, HLA-A, B, C. MHC class I antigens associated with beta 2-microglobulin are expressed by all human nucleated cells and are central in cell-mediated immune response and tumor surveillance. W6/32 mAb recognizes a non-polymorphic epitope shared among products of the HLA-A, B, and C loci and immunoprecipitates both 43kDa and 11-12kDa chains. Crossreactivity is also seen in baboon, rhesus and cynomolgus monkey.
Description & Specifications

Specifications

HLA-ABC
Murine
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
W6/32
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer