Please login to your online account to display your discounted pricing

HLA-A2 Mouse anti-Human, PE-Cy7, Clone: BB7.2, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen HLA-A2
Clone BB7.2
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123467

 
affymetrix
25987641
25 tests Each for $172.55

501129144

 
affymetrix
25987642
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen HLA-A2
Clone BB7.2
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with beta 2-microglobulin and is expressed on all nucleated cells.

The BB7.2 monoclonal antibody has been reported to exhibit blocking activity, as well as recognize HLA-A28.