Please login to your online account to display your discounted pricing

HJC 0350, Tocris™

Selective Epac2 inhibitor

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material HJC 0350
Assay Percent Range >98%
Molecular Formula C15H19NO2S
Formula Weight 277.38
Synonym 2,4-Dimethyl-1-[(2,4,6-trimethylphenyl)sulfonyl]-1H-pyrrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

484450

 
r & d systems
RDS484450
50mg Each for $746.48

484410

 
r & d systems
RDS484410
10mg Each for $177.37
Description & Specifications

Specifications

Formula Weight 277.38
Quantity 50mg
CAS 885434-70-8
Synonym 2,4-Dimethyl-1-[(2,4,6-trimethylphenyl)sulfonyl]-1H-pyrrole
Recommended Storage Store at +4°C
Chemical Name or Material HJC 0350
Assay Percent Range >98%
Molecular Formula C15H19NO2S
Safety and Handling

Store at +4°C