missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ HIV Substrate

For cellular and molecular biology applications

Specifications

Storage Requirements -30° to -10°C
Color White to off-white
Molecular Weight 1091.30
Format Lyophilized
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
Catalog Number Mfr. No. Substrate Quantity Price Quantity  
AAJ66488LB0 Alfa Aesar™
J66488LB0
5mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ66488MCR Alfa Aesar™
J66488MCR
1mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Specifications

-30° to -10°C
White to off-white
1091.30
Lyophilized