missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ HIV Envelope Protein (254-274)

For cellular and molecular biology applications

$196.42 - $299.62

Specifications

Product Type HIV Envelope Protein (254-274)
Molecular Weight 2208.56
Storage Requirements -30° to -10°C
Sequence Cys-Thr-His-Gly-Ile-Arg-Pro-Val-Val-Ser-Thr-Gln-Leu-Leu-Leu-Asn-Gly-Ser-Leu-Ala-Glu
Format Solid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66539EXD View Documents Alfa Aesar™
J66539EXD
0.5mg Each for $196.42
 
AAJ66539MCR View Documents Alfa Aesar™
J66539MCR
1mg Each for $299.62
 
Specifications

Specifications

HIV Envelope Protein (254-274)
-30° to -10°C
Solid
HPLC
2208.56
Cys-Thr-His-Gly-Ile-Arg-Pro-Val-Val-Ser-Thr-Gln-Leu-Leu-Leu-Asn-Gly-Ser-Leu-Ala-Glu
RUO

For Research Use Only