Cytiva HiTrap™ MabSelect PrismA Column PROMO
The HiTrap™ MabSelect PrismA Column is a Protein A Column.
Manufacturer:  Cytiva 29822170
Catalog No. 09928070LR2
Description
Description
  • Enhanced dynamic binding capacity compared with other protein A resins.
  • Excellent alkaline stability enables efficient cleaning and sanitization using 0.5–1.0 M NaOH.
  • Convenient HiTrap format for easy connection to a syringe, peristaltic pump, or chromatography systems such as an ÄKTA system for convenient process optimization.
Specifications
Specifications
5 mL
5 bar (0.5 MPa, 70 psi)
Antibody affinity chromatography
<20 mL/min.
1 Ea.
Column