Thermo Scientific™ HistoStar™ Embedding Workstation
Embed routine tissue samples into paraffin blocks for microtome sectioning comfortably and efficiently with the Thermo Scientific™ HistoStar™ Embedding Workstation.
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Voltage | Price | Quantity | |||||
A81000002
|
Thermo Scientific™
A81000002 |
100 to 120V |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Embed routine tissue samples into paraffin blocks for microtome sectioning comfortably and efficiently with the Thermo Scientific™ HistoStar™ Embedding Workstation. This workstation reduces fatigue and potential errors common when using other uncomfortable, hot or poorly illuminated embedding centers.
Brightly Illuminated and Clutter Free- User-adjustable LED lighting uniformly illuminates the workspace without the clutter of awkward remote lamps, reducing fatigue and minimizing errors.
- Smooth and insulated user contact points provide a cool, comfortable workspace.
- Constructed to eliminate pressure points and uncomfortable heat conditions.
- 5 liter paraffin capacity, coldplate area for 72 base molds, large heated workspace and specimen holding area enable users to manage heavy workflow in even the busiest laboratories.
- Workstation offers the first adjustable paraffin dispense paddle.
- A simple adjustment moves paddle to the most ergonomic position for each user.
- Embed even Super Mega Cassettes with ease.
- Only workstation with a heated wax trimmer built directly into the workspace.
- Remove excess paraffin at the embedding station, eliminating the need for additional equipment or wax scraping at the microtome.
- Large, easy-to-read touchscreen display allows quick access to temperature controls and other parameters.
- Programmable asleep mode saves energy during workflow lapses.
- Robust design minimizes moving parts, reducing wear and service requirements and ensuring dependable performance for years.
Specifications
50/60Hz |