Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Histone H1 Ab-1, Mouse Monoclonal Antibody

Recommended for Immunohistochemistry (Formalin/paraffin) and Immunofluorescence, Thermo Scientific™ Histone H1 Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

$462.45 - $864.86

Specifications

Antigen Histone H1 Ab-1
Clone 1415-1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS628P

 
thermo scientific™ lab vision™
MS628P
Purified Ab with BSA and Azide 1mL Each for $748.53

MS628P0

 
thermo scientific™ lab vision™
MS628P0
Purified Ab with BSA and Azide 100µL Each for $462.45

MS628P1

 
thermo scientific™ lab vision™
MS628P1
Purified Ab with BSA and Azide 500µL Each for $594.27

MS628P1ABX

 
thermo scientific™ lab vision™
MS628P1ABX
Ab Without BSA/Azide (Frozen) 100µL Each for $755.20

MS628PABX

 
thermo scientific™ lab vision™
MS628PABX
Ab Without BSA/Azide (Frozen) 200µL Each for $864.86

MS628R7

 
thermo scientific™ lab vision™
MS628R7
Ready-to-use for IHC (UVLP) 7mL Each for $777.98
Description & Specifications

Specifications

Antigen Histone H1 Ab-1
Clone 1415-1
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline DNA Repair
Type Primary
Immunogen Nuclei of myeloid leukemia biopsy cells
Isotype IgG2a

Recommended for Immunohistochemistry (Formalin/paraffin) and Immunofluorescence, Thermo Scientific™ Histone H1 Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Host Species: Mouse

Clone: 1415-1

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Nuclei of myeloid leukemia biopsy cells

Positive Control: Tonsil or lymph node

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO