Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Histone H1 Ab-1, Mouse Monoclonal Antibody


Recommended for Immunohistochemistry (Formalin/paraffin) and Immunofluorescence, Thermo Scientific™ Histone H1 Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer: thermo scientific™ lab vision™  MS628P1ABX

 View more versions of this product

Catalog No. MS628P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Isotype IgG2a
Quantity 100µL
Clone 1415-1
Format Concentrated
Antigen Histone H1 Ab-1
Host Species Mouse
Regulatory Status RUO
Cross Reactivity Human
Immunogen Nuclei of myeloid leukemia biopsy cells
Formulation Ab Without BSA/Azide (Frozen)
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Research Discipline DNA Repair
Item Description 100uL; 1mg/mL; Purified w/o BSA, azide

Recommended for Immunohistochemistry (Formalin/paraffin) and Immunofluorescence, Thermo Scientific™ Histone H1 Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Host Species: Mouse

Clone: 1415-1

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Nuclei of myeloid leukemia biopsy cells

Positive Control: Tonsil or lymph node

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not suitable)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO