Please login to your online account to display your discounted pricing

Histone Deacetylase Inhibitor II

Specifications

CAS 174664-65-4
Formula Weight 222.20g/mol
Synonym 3-Carboxy cinnamic acid bis-hydroxamide; CBHA
Storage Requirements −30° to −10°C
Format Powder
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65479LB0

 
alfa aesar™
J65479LB0
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

CAS 174664-65-4
Formula Weight 222.20g/mol
Synonym 3-Carboxy cinnamic acid bis-hydroxamide; CBHA
Storage Requirements −30° to −10°C
Format Powder
Sensitivity Light Sensitive
Description Off-white