Please login to your online account to display your discounted pricing

Alfa Aesar™ Histatin 5

For cellular and molecular biology applications

Specifications

Product Type Histatin 5
Molecular Weight 3036.29
Storage Requirements -30° to -10°C
Sequence Asp-Ser-His-Ala-Lys-Arg-His-His-Gly-Tyr-Lys-Arg-Lys-Phe-His-Glu-Lys-His-His-Ser-His-Arg-Gly-Tyr
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ64792EXD

 
alfa aesar™
J64792EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ64792MCR

 
alfa aesar™
J64792MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Histatin 5
Molecular Weight 3036.29
Storage Requirements -30° to -10°C
Sequence Asp-Ser-His-Ala-Lys-Arg-His-His-Gly-Tyr-Lys-Arg-Lys-Phe-His-Glu-Lys-His-His-Ser-His-Arg-Gly-Tyr
Format Solid
Regulatory Status RUO
Purification Method HPLC