Please login to your online account to display your discounted pricing

Histamine, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC411710050

 View more versions of this product

Catalog No. AC411710050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 111.15
Physical Form Crystalline Powder and/or Chunks
Quantity 5g
Color White to light yellow
Melting Point 81° to 85°C
Merck Index 15, 4756
Synonym 2-(4-Imidazolyl)ethylamine
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Histamine
MDL Number MFCD00005210
Molecular Formula C5H9N3
CAS 51-45-6
Packaging Glass bottle
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Contaminated work clothing should not be allowed out of the workplace.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Histamine
CAS 51-45-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H9N3
Formula Weight 111.15
MDL Number MFCD00005210
Synonym 2-(4-Imidazolyl)ethylamine
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Contaminated work clothing should not be allowed out of the workplace.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents