missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HIST3H2A, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB31311
Specifications
HIST3H2A | |
Polyclonal | |
ELISA (1:4000) Western Blot (1:1000) ChIP (0.5-1 ul/ChIP) Dot Blot (1:5000) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user. | |
histone cluster 3, H2a | |
HIST3H2A | |
A synthetic peptide (conjugated with KLH) corresponding to Histone H2A, acetylated at lysine 5. | |
RUO | |
92815 | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
In PBS (0.05% sodium azide, 0.05% proclin 300). | |
MGC3165 | |
Rabbit | |
50 μg | |
Primary | |
Human | |
Antibody |