Please login to your online account to display your discounted pricing

GE Healthcare HiScreen™ Capto™ S, Capto™ Q and Capto™ DEAE Prepacked IEX Columns

Use these columns for method optimization and parameter screening. GE Healthcare HiScreen™ Capto™ S, Capto™ Q and Capto™ DEAE Prepacked IEX Columns combine high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity.

$147.53 - $147.53

Specifications

Buffer 1M Acetic Acid, 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Bed Volume 4.7mL
Diameter (Metric) Inner 7.7mm
Height (Metric) 100mm
Flow Rate <700cm/hr.
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

45002529PM

 
ge healthcare
28926978PM
HiScreen Capto Q IEX column Each for $147.53

45002530PM

 
ge healthcare
28926979PM
HiScreen Capto S IEX column Each for $147.53
Description & Specifications

Specifications

Buffer 1M Acetic Acid, 1M NaOH, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol and 70% Ethanol
Bed Volume 4.7mL
Diameter (Metric) Inner 7.7mm
Height (Metric) 100mm
Flow Rate <700cm/hr.
Max. Pressure 3 bar
Storage Requirements 4° to 30°C
Material Polypropylene

  • Fast, efficient, and cost-effective capture and intermediate protein purification
  • Ready-to-use columns for convenient process development optimization
  • Highly cross-linked agarose with dextran surface extender matrix
  • Bed height of 10cm is designed to allow method optimization and parameter screening
  • Easily connected in series to achieve 20cm bed height
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Combines high capacity with high flow velocity and low backpressure to reduce process cycle times and increase productivity
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

Capto S: Strong cation (sulfonate group) exchange BioProcess™ medium
Capto Q: Strong anion (quartenary amine group) exchange BioProcess medium
Capto DEAE: Weak anion (diethylaminoethyl group) exchange BioProcess medium