Please login to your online account to display your discounted pricing

His Tag Mouse anti-Mouse, Clone: HIS.H8, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $366.85

Specifications

Antigen His Tag
Clone HIS.H8
Host Species Murine
Gene Alias 6X His tag
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122712

 
life technologies
LS14665780
25μg Each for $160.95

501122713

 
life technologies
LS14665782
100μg Each for $366.85
Description & Specifications

Specifications

Antigen His Tag
Clone HIS.H8
Host Species Murine
Gene Alias 6X His tag
Species Reactivity Mouse
Applications Immunoprecipitation
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This HIS.H8 monoclonal antibody reacts with the 6X polyhistidine (6X His) tag. The H-H-H-H-H-H motif is frequently fused to the N- or C-terminus of proteins to facilitate purification (e.g., nickel affinity chromatography and immunoprecipitation) and/or detection. This antibody recognizes the 6X His tag anywhere within the protein of interest (i.e., N-terminus, C-terminus, or internal).