Please login to your online account to display your discounted pricing

Alfa Aesar™ [His&superscript_7;] Corazonin

For cellular and molecular biology applications

Specifications

Product Type [His7] Corazonin
Molecular Weight 1350.41
Storage Requirements -30° to -10°C
Sequence Glp-Thr-Phe-Gln-Tyr-Ser-His-Gly-Trp-Thr-Asn-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66315LB0

 
alfa aesar™
J66315LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66315MCR

 
alfa aesar™
J66315MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [His7] Corazonin
Molecular Weight 1350.41
Storage Requirements -30° to -10°C
Sequence Glp-Thr-Phe-Gln-Tyr-Ser-His-Gly-Trp-Thr-Asn-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC