Learn More
Hippuric acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1269036
Description
Hippuric acid is used as a intermediate for the manufacturing medicine and other organic compounds. Hippuric acid can be used to study cell biology, chemical biology, bioactive small molecules, amino acid derivatives, peptide synthesis, chemical synthesis and nutrition. Hippuric acid has been used to inform the metabolism and urinary excretion of procyanidins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHippuric acid is used as a intermediate for the manufacturing medicine and other organic compounds. Hippuric acid can be used to study cell biology, chemical biology, bioactive small molecules, amino acid derivatives, peptide synthesis, chemical synthesis and nutrition. Hippuric acid has been used to inform the metabolism and urinary excretion of procyanidins.
Solubility
Soluble in water.
Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
Hippuric acid | |
188°C to 191°C | |
C9H9NO3 | |
MFCD00002692 | |
1073987 | |
hippuric acid, n-benzoylglycine, glycine, n-benzoyl, benzoylglycine, benzamidoacetic acid, benzoylaminoacetic acid, hippurate, hippursaeure, phenylcarbonylaminoacetic acid, benzamidoessigsaeure | |
QIAFMBKCNZACKA-UHFFFAOYSA-N | |
2-benzamidoacetic acid | |
464 | |
179.18 |
495-69-2 | |
1.371 | |
C6H5CONHCH2CO2H | |
500 g | |
14,4716 | |
Soluble in water. | |
OC(=O)CNC(=O)C1=CC=CC=C1 | |
179.18 | |
CHEBI:18089 | |
98% |
Chemical Identifiers
495-69-2 | |
179.18 | |
QIAFMBKCNZACKA-UHFFFAOYSA-N | |
464 | |
2-benzamidoacetic acid |
C9H9NO3 | |
MFCD00002692 | |
hippuric acid, n-benzoylglycine, glycine, n-benzoyl, benzoylglycine, benzamidoacetic acid, benzoylaminoacetic acid, hippurate, hippursaeure, phenylcarbonylaminoacetic acid, benzamidoessigsaeure | |
CHEBI:18089 | |
OC(=O)CNC(=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
EINECSNumber : 207-806-3
RTECSNumber : MR8150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only