missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ HIPK2 Recombinant Human Protein DFS Item

HIPK2 is a nuclear serine/threonine kinase.

$774.00 - $11260.00

Specifications

Accession Number NP_073577.3
Molecular Weight (g/mol) 73kDa
Storage Requirements Kinase proteins are shipped on dry ice and should be stored at -80°C.
Gene Alias HIPK2
KinaseFamily DYRK Kinase Family
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV5275 DFS Item
View Documents Promotion Details
Invitrogen™
PV5275
10μg
Each for $774.00
 
PR9004A DFS Item
View Documents Promotion Details
Invitrogen™
PR9004A
100μg
Each for $2,650.00
 
PV5276 DFS Item
View Documents Promotion Details
Invitrogen™
PV5276
1mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

HIPK2 is linked to transcriptional regulation and apoptosis.
Specifications

Specifications

NP_073577.3
Kinase proteins are shipped on dry ice and should be stored at -80°C.
DYRK Kinase Family
Recombinant
Baculovirus
73kDa
HIPK2
Human
GST Tag
SDS
missing translation for 'documents'

missing translation for 'documents'